Skip to Content
Merck
All Photos(1)

Key Documents

C71203

Sigma-Aldrich

4-Chlorostyrene

97%, contains 500 ppm 4-tert-butylcatechol as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4Cl
CAS Number:
Molecular Weight:
138.59
Beilstein:
1098859
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.68 mmHg ( 20 °C)

Quality Level

Assay

97%

form

liquid

contains

500 ppm 4-tert-butylcatechol as inhibitor

refractive index

n20/D 1.565 (lit.)

bp

192 °C (lit.)

density

1.155 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1ccc(C=C)cc1

InChI

1S/C8H7Cl/c1-2-7-3-5-8(9)6-4-7/h2-6H,1H2

InChI key

KTZVZZJJVJQZHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chlorostyrene is a para-halogenated styrene derivative. It undergoes graft copolymerization with acrylonitrile (AN) onto ethylene-propylene-diene terpolymer (EPDM) in the presence of benzoyl peroxide (initiator).

Application

4-Chlorostyrene was used in the following studies:
  • Study of chemical and biochemical properties of the vinyl group of styrene by the development of structure activity relationships (SAR).
  • Preparation of new bis(pyrazolyl)borato olefin complexes of copper(I).
  • To investigate the regioselectivity in the cationic Heck reaction of 4-substituted styrenes.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guido Pampaloni et al.
Dalton transactions (Cambridge, England : 2003), (29)(29), 3576-3583 (2006-07-21)
New bis(pyrazolyl)borato olefin complexes of copper(I) of general formula Cu[BH2(3,5-(CF3)2Pz)2](olefin) have been prepared (olefins: coe = cyclooctene, van = 4-vinylanisole, clsty = 4-chlorostyrene, tevs = triethylvinylsilane, fn = fumaronitrile). The structures of Cu[BH2(3,5-(CF3)2Pz)2](L), L = coe, van, tevs, fn, have
Wei Chen et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 29(2), 152-153 (2011-05-31)
To establish a rapid gas chromatographic method for determination of o-chlorostyrene in the air of workplace. The air samples were collected by syringes, injected directly to the GC system, and then separated by a FFAP capillary column (30 m ×
Reactivity and regioselectivity in the Heck reaction: Hammett study of 4-substituted styrenes.
Fristrup P, et al.
Organometallics, 23(26), 6160-6165 (2004)
Synthesis and properties of acrylonitrile-EPDM-4-chlorostyrene graft copolymer.
Park D-J, et al.
Journal of Applied Polymer Science, 44(4), 727-735 (1992)
Małgorzata Bołt et al.
Materials (Basel, Switzerland), 13(18) (2020-09-12)
Bifunctional silsesquioxanes create an attractive group of compounds with a wide range of potential applications, and recently they have gained much interest. They are known to be obtained mainly via hydrosilylation, but we disclose novel synthetic protocols based on different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service