C30604
4-Chlorobutyryl chloride
99%
Synonym(s):
4-Chlorobutanoic acid chloride, 4-Chlorobutyric acid chloride
About This Item
Recommended Products
vapor pressure
3 mmHg ( 20 °C)
Quality Level
Assay
99%
form
liquid
autoignition temp.
824 °F
refractive index
n20/D 1.461 (lit.)
bp
173-174 °C (lit.)
density
1.26 g/mL at 25 °C (lit.)
SMILES string
ClCCCC(Cl)=O
InChI
1S/C4H6Cl2O/c5-3-1-2-4(6)7/h1-3H2
InChI key
CDIIZULDSLKBKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 1
Flash Point(F)
185.0 °F
Flash Point(C)
85 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service