Skip to Content
Merck
All Photos(2)

Key Documents

C101206

Sigma-Aldrich

1,4-Cyclohexanediol

99%

Synonym(s):

Hexahydrohydroquinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(OH)2
CAS Number:
Molecular Weight:
116.16
Beilstein:
2036658
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

bp

150 °C/20 mmHg (lit.)

mp

98-100 °C (lit.)

SMILES string

OC1CCC(O)CC1

InChI

1S/C6H12O2/c7-5-1-2-6(8)4-3-5/h5-8H,1-4H2

InChI key

VKONPUDBRVKQLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Wu et al.
Applied microbiology and biotechnology, 73(5), 1097-1105 (2006-09-16)
Commercial formulations of hexachlorocyclohexane (HCH) consist of a mixture of four isomers: alpha, beta, gamma, and delta. All four isomers are toxic and recalcitrant pollutants. beta-HCH is more problematic due to its longer persistence in the environment. Sphingomonas sp. BHC-A
Shuai Shuai Wu et al.
Cryo letters, 34(5), 497-507 (2014-01-23)
In this study, the fibroblasts cell line derived from ear marginal tissue of Yunnan semi-fine wool sheep was successfully established using the primary explants technique and cryopreservation technology. Additionally, the protective effect of synthetic ice blocker (SIB) including 1, 3-cyclohexanediol
G Giavaresi et al.
The International journal of artificial organs, 27(9), 796-805 (2004-11-04)
Two natural Biopol polyesters, containing 8% (D400G) and 12% (D600G) of hydroxyvalerate component, and a synthetic polyester based on 1,4 cyclohexanediol [Poly(cyclohexyl-sebacate)--PCS] were studied to investigate their in vitro and in vivo behavior for application in musculoskeletal tissue repair. The
James H Loehlin et al.
Acta crystallographica. Section B, Structural science, 64(Pt 5), 583-588 (2008-09-19)
The structure of a co-crystal with both trans and cis isomers of 1,4-cyclohexanediol (1,4-CHD) is reported. The intermolecular hydrogen-bond patterns are described and compared with those of the all trans structure, using the graph-set model. A second crystal with possible
Stéphane Patry et al.
Carbohydrate research, 500, 108217-108217 (2020-12-16)
In this paper, the synthesis of different bisglucosides is investigated through the reaction of two acetylated glucose units with a diol (or diphenol) in order to develop a versatile molecular platform for the future development of bio-based polymers. A panel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service