BR90617
BRAND® glass beaker with spout, tall form
volume 50 mL, with graduation
Synonym(s):
Beakers, Glass
All Photos(2)
About This Item
Recommended Products
material
borosilicate glass 3.3
feature
with graduation
manufacturer/tradename
BRAND 90617
volume
50 mL
Related Categories
General description
The BRAND® Glass Beaker with Spout, tall form, is constructed from high-quality Boro 3.3 glass, ensuring durability and reliability in laboratory environments. Compliant with DIN EN ISO 3819 standards, this beaker features clear graduation markings for accurate liquid measurement. The tall form design provides increased capacity, while the integrated spout facilitates easy pouring, minimizing spills and enhancing precision during liquid transfer.
Other Notes
Browse all glass beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)