BR759215
BRAND® UV cuvette micro
center H 8.5 mm, volume 70-850 μL, pack of 100 ea (individually wrapped)
Synonym(s):
Cuvettes, UV-Cuvettes
All Photos(1)
About This Item
UNSPSC Code:
41121813
NACRES:
SB.32
Recommended Products
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
BRAND 759215
center H
8.5 mm
volume
70-850 μL
Related Categories
General description
- BRAND® UV cuvette micro is manufactured under controlled room conditions.
- Ideal for the determinations in water analysis, chemistry, and life science applications.
- Cuvette material offers high chemical resistance to most polar solvents, acids, and alkaline solutions.
- Clear, clean optical path with an indication of optical path orientation.
- Eliminates the risk of cross-contamination.
- Grouped by mold cavity number.
- An economical alternative to Quartz cuvettes.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)