Key Documents
BR718605
BRAND® counting chamber BLAUBRAND® Neubauer pattern
without clips, double ruled
Synonym(s):
BLAUBRAND®, Cell culture, Counting Chambers, Neubauer, ldpe bottle, polyethylene bottles, polyethylene containers
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Pricing and availability is not currently available.
material
colorless
glass
sterility
non-sterile
feature
without clips
manufacturer/tradename
BRAND 718605
chamber size
0.1 mm , depth
wells
1
General description
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The ruling shows 9 large squares of 1 mm² each.The four large squares in the corners marked "L" for counting leucocytes are each subdivided into 16 squares with 0.25 mm sides.The large square in the center is subdivided into 16 squares with 0.25 mm sides. Each group square consists of 16 mini squares with 0.05 mm sides, each having an area of 0.0025 mm². The 5 group squares in the large central square are used for counting thrombocytes and erythrocytes.Contrary to the more advanced Improved Neubauer counting chambers, the counting area of each group square is limited by the outer one of the triple boundary lines.
Features and Benefits
• BLAUBRAND® quality• Outstanding measurement precision• 100% certified quality
Packaging
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)