BR708012
Discharge tube for Dispensette® S
Organic, capacity 5, 10 mL, fine tip
, L 105 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.11
material
polypropylene cap
feature
Organic
manufacturer/tradename
BRAND 708012
L
105 mm
capacity
5, 10 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Without recirculation valve
Legal Information
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service