BR701623
BRAND® Seal-R-film™ Sealing Film
roll L × W 75 m (50) mm, Single Roll
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
30102015
NACRES:
NB.35
Recommended Products
form
film
packaging
case of 1 ea
manufacturer/tradename
BRAND 701623
roll L × W
75 m (50) mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
No more spilling, evaporation, and contamination! Seal-R-film™ sealing film seals beakers, flasks, and other vessels.
The sealing film can be stretched to three times its original length. This means that Seal-R-film™ always fits, even with irregular shapes. For traceability, each roll has a lot identification. With the BRAND cutter for sealing film you can cut rolls with widths of 50 and 100 mm efficiently and economically and store them cleanly.
The sealing film can be stretched to three times its original length. This means that Seal-R-film™ always fits, even with irregular shapes. For traceability, each roll has a lot identification. With the BRAND cutter for sealing film you can cut rolls with widths of 50 and 100 mm efficiently and economically and store them cleanly.
Features and Benefits
- Protects against evaporation, leakage, and contamination
- Easy sealing with 300% elasticity
- Lot identification of each roll
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Seal-R-film is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service