Key Documents
BR472600
BRAND® glass staining trough, Hellendahl pattern
soda-lime glass
Synonym(s):
Microscopy, Staining, Staining Accessories
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
material
soda-lime glass
manufacturer/tradename
BRAND 472600
L × W
76 mm × 26 mm , holds 16 slides
General description
The BRAND® Glass Staining Trough in the Hellendahl pattern is specifically designed for the efficient staining of up to 16 slides, making it an essential tool for histology and microscopy applications. Constructed from durable soda-lime glass, this trough measures 76 x 26 mm, providing ample space for organizing multiple slides. The Hellendahl design ensures optimal placement and accessibility, facilitating uniform staining and enhancing laboratory workflow. This reliable and practical staining trough is a valuable addition to any laboratory setup, ensuring effective and efficient staining processes.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)