BR4600100
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 0.1-1 mL, without recirculation valve
Synonym(s):
bottle top dispensers
About This Item
Recommended Products
feature
incremental 0.02 mL
DE-M marking
without recirculation valve
manufacturer/tradename
BRAND 4600100
volume range
0.1-1 mL
fitting
(40 mm)
(GL 24-25)
GL 32
GL 38
S 40
Related Categories
General description
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Protocols
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Related Content
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.