BR37491
BRAND® volumetric flask BLAUBRAND® USP
capacity 1000 mL, joint: 24/29, polypropylene stopper
Synonym(s):
BLAUBRAND®, USP, Volumetric flasks
About This Item
Recommended Products
material
amber
borosilicate glass 3.3 flask
polypropylene stopper
Joint
joint ST/NS neck24/29
joint: 24/29
feature
class A DE-M marking. To DIN EN ISO 1042. Calibrated to contain (TC, In). With USP batch certificate.
DE-M marking
packaging
pack of 1 ea
manufacturer/tradename
BRAND 37491
capacity
1000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service