BR23830
BRAND® compact automatic SILBERBRAND burette, without support/clamp
volume 50 mL, with LDPE bottle and PVC filling tube
Synonym(s):
Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
All Photos(2)
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
Recommended Products
material
borosilicate glass (5.4)
manufacturer/tradename
BRAND 23830
volume measuring range
50 mL, accuracy: ±0.10 mL
volume
50 mL
Related Categories
General description
The Modular Automatic Burette Concept! Fast to dismantle and easy to clean. Easy to repair - all individual components replaceable! Borosilicate glass 5.4, detachable PTFE stopcock. Calibrated to deliver (TD, Ex). 25 ml: Error limit according to class B, DIN EN ISO 385. 50 ml: DIN EN ISO 385, class B.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)