BR152907
BRAND® tubing connector, Y-shape
polypropylene, for tubing i.d., 8 ‑ 9 mm
Synonym(s):
Tubing, Tubing Connectors
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polypropylene
feature
for tubing i.d. 8 ‑ 9 mm
packaging
pack of 20 ea
manufacturer/tradename
BRAND 152907
L
64 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The BRAND® Y-shaped tubing connector is made from translucent polypropylene (PP), providing clear visibility of fluid flow. This connector is designed to create a branching point in tubing systems, allowing for the connection of multiple tubes at a Y junction. Its durable construction ensures a secure and reliable fit, reducing the risk of leaks or disconnections during use.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service