Key Documents
BR150907
BRAND® watch glass
PTFE, diam. 65 mm
Synonym(s):
Dishes, PTFE, Watch glasses
About This Item
UNSPSC Code:
41121801
NACRES:
NB.43
material
PTFE
manufacturer/tradename
BRAND 150907
diam.
65 mm
General description
The BRAND® watch glass is made from high-quality PTFE, making it ideal for covering PTFE beakers. Its excellent chemical resistance ensures durability and reliability when handling a wide range of substances. This watch glass provides a secure cover to prevent contamination and evaporation during experiments. Its lightweight and non-reactive properties make it a valuable accessory in laboratory settings, enhancing the efficiency of various applications. This versatile tool is essential for maintaining the integrity of samples and solutions in the lab.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)