BR150015
BRAND® watch glass
soda-lime glass, diam. 50 mm , ground edges stress-reduced
Synonym(s):
Dishes, Watch glasses
All Photos(2)
About This Item
Recommended Products
material
soda-lime glass
manufacturer/tradename
BRAND 150015
diam.
50 mm , ground edges stress-reduced
Related Categories
General description
The BRAND® watch glass is made from soda-lime glass and complies with DIN 12341 standards. It features ground edges for safe handling and a refined finish. This watch glass is stress-reduced to minimize the risk of shattering, enhancing safety during laboratory use. Ideal for covering beakers or evaporating dishes, it provides an effective solution for preventing contamination and evaporation of samples. Its durable design makes it a reliable tool in various laboratory applications, ensuring optimal performance in your experiments.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)