BR145505
BRAND® funnel short stem, glass
short stem, borosilicate glass, diam. 35 mm
Synonym(s):
Funnels
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
feature
for rapid filtration
manufacturer/tradename
BRAND 145505
diam.
35 mm
stem diam. × L
6 mm × 35 mm
General description
The BRAND® Funnel with Short Stem is designed for efficient and rapid filtration in laboratory applications. Made from high-quality Boro 3.3 glass, this funnel combines durability with excellent performance. The short stem facilitates quick liquid transfer and minimizes the risk of clogging during filtration processes. The funnel is angled at 60°, optimizing flow rates. The upper rim is either ground or fire-polished, ensuring a smooth finish that enhances safety and ease of use.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)