BR115510
BRAND® test tube rack
PTFE, diam. 13 mm , 21 tubes
Synonym(s):
Accessories, Test Tube Racks, Tubes
About This Item
Recommended Products
material
PTFE
manufacturer/tradename
BRAND 115510
L × W × H
180 mm × 60 mm × 60 mm
diam.
13 mm , 21 tubes
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- BRAND® test tube racks are sturdy with excellent mechanical strength.
- Chemically resistant to most reagents.
- Suitable for use in temperatures from -200 °C to 250 °C.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service