Key Documents
BR114820
BRAND® PP graduated centrifuge tube, screw cap
volume 50 mL, suitable for PFAS testing, without base, non-sterile
Synonym(s):
Centrifuge tubes, Tubes
About This Item
UNSPSC Code:
41121703
NACRES:
NB.43
Pricing and availability is not currently available.
material
polypropylene
screw top cap
translucent tube
sterility
non-sterile
feature
RCF 3,000 × g
without base
packaging
pack of 300
manufacturer/tradename
BRAND 114820
volume
50 mL
General description
The BRAND® PP Graduated Centrifuge Tube with Screw Cap is made from high-quality polypropylene (PP) and features a translucent design for easy visibility of contents. This tube is engineered to withstand relative centrifugal forces (RCF) of up to 3000, making it suitable for various laboratory applications. It comes graduated, allowing for precise measurements, and is available in both non-sterile and gamma-ray sterilized options in 15 ml (graduated up to 13 ml) or 50 ml capacities. The screw cap, made from polyethylene (PE) and equipped with a sealing cone, ensures a secure closure to prevent leaks. This centrifuge tube offers high clarity and reliability, making it an essential tool for researchers and technicians in the lab.
Features and Benefits
  • BRAND® graduated centrifuge tube is the ideal solution for all your centrifuge needs.
  • Made from highly transparent polypropylene (PP) for enhanced sample visibility.
  • Bio-inert PP offers optimal cell and protein recovery.
  • Includes a screw cap with a plug for preventing sample loss.
  • Tubes are tested to withstand RCF up to 3000.
  • γ-ray sterilized.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Rapid & accurate analysis of 208 pesticides and their metabolites in a fruit sample using Supel™ QuE QuEChERS mixes for sample preparation and GC-MS/MS.
Rapid & accurate analysis of 208 pesticides and their metabolites in a fruit sample using Supel™ QuE QuEChERS mixes for sample preparation and GC-MS/MS.
Rapid & accurate analysis of 208 pesticides and their metabolites in a fruit sample using Supel™ QuE QuEChERS mixes for sample preparation and GC-MS/MS.
Rapid & accurate analysis of 208 pesticides and their metabolites in a fruit sample using Supel™ QuE QuEChERS mixes for sample preparation and GC-MS/MS.
See All