BMSE4000EUK
Benchmark SmartBlue Transilluminator
AC/DC input 230 V, UK plug
Synonym(s):
SYBR, SmartGlow, blue light, gel imaging, gel casting, transillumination
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Benchmark Scientific E4000-E-UK
Looking for similar products? Visit Product Comparison Guide
General description
Accuris SmartBlue Transilluminators have been developed for viewing DNA gels stained with new, safe fluorescent dyes. These new dyes, such as SmartGlow, SYBR Green, Gel Green, have an excitation wavelength of 465470nm (visible blue light), and are safer to use than Ethidium Bromide.
Application
Electrophoresis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service