Key Documents
BAH942404004
Scienceware® Lab Companion perforated shelf
fits 11 L cabinet vacuum desiccators, pack of 1 ea
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H94240-4004
General description
SP Bel-Art Perforated Shelf, available as a replacement of or addition to Lab Companion™ cabinet style vacuum desiccators. Perforated shelves permit efficient airflow throughout cabinet; all models allow for additional shelves to be added.
Legal Information
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)