BAF378960000
SP Bel-Art® Pipette Pump
capacity 0.2 mL, yellow
Synonym(s):
Pump For Pipette
All Photos(2)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
Recommended Products
material
yellow
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F37896-0000
capacity
0.2 mL
Related Categories
General description
Provides safe, accurate and trouble-free pipetting with simple, one-handed operation.
Features and Benefits
- Pipettes fit smoothly into its flexible tapered chuck
- Thumbwheel on side is rotated for precision aspirating or dispensing
- Plunger may be pressed for quick emptying
- Resistant to acids and alkalis
- Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
- Replacement Silicone Rubber Chuck and Washers are available
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)