BAF199170002
Scienceware® vacuum aspirator bottle with trap
capacity 1.0 gal
Synonym(s):
bottle aspirator
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.79
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F19917-0002
capacity
1.0 gal
Related Categories
General description
Make Quick Work of Large Jobs
For easy and continuous aseptic vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes. To use, simply insert a pipette tip, Pasteur pipette or serological pipette into the collection tube and connect vacuum line to vacuum source.
For easy and continuous aseptic vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes. To use, simply insert a pipette tip, Pasteur pipette or serological pipette into the collection tube and connect vacuum line to vacuum source.
- Chemical and corrosion resistant polypropylene bottle and epoxy coated steel frame
- Will not crowd workspace, fume hood, or laminar flow hood
- Quickly and conveniently aspirate and collect fluids using a vacuum source
- Holds 3200 mL (108 oz) and the included trap prevents fluid flow into the vacuum source
- Use to aspirate media, waste chemicals, cell washing solutions, and more
- Ideal for use with dishes, plates, flasks and tubes
- All parts, except the clear PVC tubing, are autoclavable at 121 °C (250 °F)
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)