Key Documents
BAF182350000
Tubing holder clip for beakers
pack of 6 ea
About This Item
UNSPSC Code:
41121803
NACRES:
NB.51
material
polypropylene
feature
autoclavable
packaging
pack of 6 ea
manufacturer/tradename
Bel-Art F18235-0000
Related Categories
General description
This polypropylene clip captures and holds flexible tubing firmly in place in beakers or other open containers. It fits snugly over the edge and has a 51 mm (2 in.) long arm with a hole to accept tubing up to 9.5 mm (3/8 in.) O.D. 6 per bag
Holds tubing of 3/8 in. OD in place over edge of beakers or other open containers with a wall thickness of up to 1/4 in.
Choose from one of the most recent versions:
Certificates of Analysis (COA)