Skip to Content
Merck
All Photos(1)

Key Documents

B5136

Sigma-Aldrich

Benzylamine hydrochloride

Synonym(s):

Phenylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NH2·HCl
CAS Number:
Molecular Weight:
143.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

crystals

Quality Level

mp

262-263 °C (lit.)

SMILES string

Cl.NCc1ccccc1

InChI

1S/C7H9N.ClH/c8-6-7-4-2-1-3-5-7;/h1-5H,6,8H2;1H

InChI key

XKXHCNPAFAXVRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syed Suhail Andrabi et al.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer, 39(1), 1-12 (2020-06-02)
Glutamatergic N-methyl-D-aspartate (NMDA) receptors have critical roles in several neurological and psychiatric diseases. Dizocilpine (MK-801) is a ligand at phencyclidine recognition sites that is associated with NMDA receptor-coupled cation channels, where it acts as a potent noncompetitive antagonist of central
Ibrahim Olabayode Saliu et al.
Journal of ethnopharmacology, 264, 113284-113284 (2020-08-26)
Tetrapleura tetraptera Taub. (family Fabaceae), is generally found in the lowland forest of tropical Africa. Its leaves and fruits are traditionally used in West Africa for the management of brain disorders. This study evaluated the effect of Tetrapleura tetraptera methanol
Min-Ho Nam et al.
ACS chemical neuroscience, 8(7), 1519-1529 (2017-03-24)
To develop novel, selective, and reversible MAO-B inhibitors for safer treatment of Parkinson's disease, benzothiazole and benzoxazole derivatives with indole moiety were designed and synthesized. Most of the synthesized compounds showed inhibitory activities against MAO-B and selectivity over MAO-A. The
Saioa Gomez-Zorita et al.
Journal of physiology and biochemistry, 69(3), 585-593 (2013-01-15)
Resveratrol is a naturally occurring polyphenol found in many dietary sources and red wine. Recognized as a cancer chemoprevention agent, an anti-inflammatory factor and an antioxidant molecule, resveratrol has been proposed as a potential anti-obesity compound and to be beneficial
Danielle M Shacklady-McAtee et al.
Organic letters, 13(13), 3490-3493 (2011-06-08)
A nickel(0) catalyst effectively mediates the cyclization of N-benzoyl aminals in the presence of a stoichiometric Lewis acid. This method enables preparation of a variety of isoindolinones with substitution on the benzoyl fragment and C-3 carbon. This reaction likely proceeds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service