Skip to Content
Merck
All Photos(1)

Key Documents

ALD00600

Sigma-Aldrich

Zhu-Yu Auxiliary

98%

Synonym(s):

1-Carboxy-1-methylethoxyammonium chloride, 2,2-Dimethyl aminooxyacetic, 2-(Aminooxy)-2-methylpropanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10ClNO3
CAS Number:
Molecular Weight:
155.58
MDL number:
UNSPSC Code:
12352101

Assay

98%

form

powder or crystals

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

availability

available only in USA

mp

150-165 °C

InChI

1S/C4H9NO3.ClH/c1-4(2,8-5)3(6)7;/h5H2,1-2H3,(H,6,7);1H

InChI key

ZJPZGAAOCIFBDI-UHFFFAOYSA-N

Application

The Zhu-Yu Auxiliary was reported by Jin-Quan Yu′s lab to be an effective auxiliary for the Pd(II)-catalyzed ß-C(sp3)-H(hetero)arylation of ketones.

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Yu program centers around the discovery of catalytic carbon–carbon and carbon–heteroatom bond forming reactions based on C–H activation. Target transformations are selected to enable 1) the use of simple and abundant starting materials such as aliphatic acids, amines and alcohols, and 2) disconnections that drastically shorten the synthesis of a drug molecule or a major class of biologically active compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service