Key Documents
ACE585326
Ace-Safe® tubing connector, UHDPE
#15 to 1/2 in. I.D. tubing
About This Item
UNSPSC Code:
41106514
NACRES:
NB.79
packaging
pack of 1 ea
manufacturer/tradename
Ace Glass 5853-26
parameter
-20-110 °C temp. limit
Related Categories
General description
Tubing connector, used to connect flexible tubing (1/4 in., 3/8 in., 1/2 in., 3/4 in., 1 in. I.D.) to #7, #11, #15 or #25 Ace-Thred for easy, safe connect/disconnect. Nylon bushing slides over serrated end and secures PP connector in thread with silicone O-ring in front groove to make compression seal. With tubing attached at serrated end, un-thread bushing and connector is removed from thread.
Legal Information
Ace-Safe is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)