A79604
4-Aminosalicylic acid
99%, for peptide synthesis
Synonym(s):
4-Amino-2-hydroxybenzoic acid, PAS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Beilstein:
473071
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Product Name
4-Aminosalicylic acid, 99%
Assay
99%
form
powder
reaction suitability
reaction type: solution phase peptide synthesis
color
white to beige
mp
135-145 °C (lit.)
application(s)
peptide synthesis
SMILES string
Nc1ccc(C(O)=O)c(O)c1
InChI
1S/C7H7NO3/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,9H,8H2,(H,10,11)
InChI key
WUBBRNOQWQTFEX-UHFFFAOYSA-N
Gene Information
human ... PTPN1(5770)
Looking for similar products? Visit Product Comparison Guide
Application
4-Aminosalicylic acid (4-ASA) can be used in the synthesis of:
- Azo derivatives of 4-ASA with anti-inflammatory effects.
- Ammonium 4-aminosalicylate salt polymorphs which are used as pharmaceutical ingredients.
- Salicylic acid-triazole analogs which are used as quorum sensing inhibitors against Pseudomonas aeruginosa.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of azo derivatives of 4-aminosalicylic acid.
Zhao ZB, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 18(6), 639-642 (2007)
Design, Synthesis and Biological Evaluation of Triazole-Containing 2-Phenylindole and Salicylic Acid as Quorum Sensing Inhibitors Against Pseudomonas aeruginosa.
Srinivasarao S, et al.
ChemistrySelect, 3(32), 9170-9180 (2018)
Polymorphic ammonium salts of the antibiotic 4-aminosalicylic acid.
Andre V, et al.
Crystal Growth & Design, 12(6), 3082-3090 (2012)
Tian-Loon Lee et al.
Journal of glaucoma, 23(9), 644-648 (2013-02-23)
To determine the incidence of intraocular pressure (IOP) spikes within the first 30 minutes after sequential argon-Nd:YAG laser peripheral iridotomy (LPI) in patients with angle closure and to explore risk factors for their occurrence. A total of 428 consecutive eyes
Maria João Forjaz et al.
Movement disorders : official journal of the Movement Disorder Society, 30(4), 545-551 (2014-12-30)
The Parkinson Anxiety Scale is a new scale developed to measure anxiety severity in Parkinson's disease specifically. It consists of three dimensions: persistent anxiety, episodic anxiety, and avoidance behavior. This study aimed to assess the measurement properties of the scale
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service