Skip to Content
Merck
All Photos(3)

Key Documents

A75633

Sigma-Aldrich

2-Amino-3-methylpyridine

95%

Synonym(s):

2-Amino-3-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
107892
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

refractive index

n20/D 1.5823 (lit.)

bp

221-222 °C (lit.)

mp

29-31 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

Cc1cccnc1N

InChI

1S/C6H8N2/c1-5-3-2-4-8-6(5)7/h2-4H,1H3,(H2,7,8)

InChI key

RGDQRXPEZUNWHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T G Altuntas et al.
Anticancer research, 17(5A), 3485-3491 (1997-12-31)
The metabolism of the comutagen 2-amino-3-methylpyridine has been studied in vitro using rat and rabbit hepatic preparations. 2-Amino-3-methylpyridine-N-oxide, 2-amino-3-hydroxymethylpyridine and 2-amino-5-hydroxy-3-methylpyridine were formed by both rat and rabbit hepatic preparations. No evidence was obtained for the formation of the corresponding
T G Altuntas et al.
Anticancer research, 17(6D), 4479-4482 (1998-03-12)
The metabolism of 2-amino-3-methylpyridine (2A3MP) in vitro has been investigated using the rat, rabbit, dog, marmoset, guinea pig and hamster hepatic microsomes and S9 supernatants (10,000 g fraction). Species differences were observed in the in vitro formation of 2-amino-3-methylpyridine-N-oxide (2A3MPNO)
T G Altuntas et al.
Xenobiotica; the fate of foreign compounds in biological systems, 27(11), 1103-1111 (1997-12-31)
1. The effects of various potential inhibitors, activators and inducers on the metabolism of the comutagen 2-amino-3-methylpyridine (2A3MP) by rabbit hepatic microsomes and S9 supernatants have been studied. 2. The 1-N-oxidation of 2A3MP to 2-amino-3-methylpyridine-1-N-oxide (2A3M-PNO) was inhibited by 2,4-dichloro-6-phenylphenoxyethylamine
Cengiz Yenikaya et al.
Journal of enzyme inhibition and medicinal chemistry, 26(1), 104-114 (2010-09-24)
Two novel proton transfer compounds were prepared between 2,4-dichloro-5-sulphamoylbenzoic acid (lasamide) (Hsba) and ethylenediamine (en), namely ethane-1,2-diaminium 2,4-dichloro-5-sulphamoylbenzoate (1), and also between Hsba and 2-amino-3-methylpyridine (2-amino-3-picoline) (amp), namely 2-amino-3-methylpyridinium 2,4-dichloro-5-sulphamoylbenzoate (2). All these were characterised by elemental, spectral (IR and
J W Gorrod et al.
European journal of drug metabolism and pharmacokinetics, 22(4), 341-344 (1998-03-26)
The in vitro metabolism of 2-hydroxylamino-3-methylpyridine has been investigated using arochlor 1254 pretreated rat S9 mixtures. 2-Hydroxylamino-3-methylpyridine is rapidly converted to the parent amine 2-amino-3-methylpyridine. No further oxidation products of 2-hydroxylamino-3-methylpyridine (i.e. nitroso or nitro) were detected under the HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service