Skip to Content
Merck
All Photos(1)

Key Documents

93690

Sigma-Aldrich

L-Tryptophan ethyl ester hydrochloride

≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O2 · HCl
CAS Number:
Molecular Weight:
268.74
Beilstein:
3919010
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (AT)

form

powder

optical activity

[α]20/D +10±1°, c = 2% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

220-225 °C (dec.)

application(s)

peptide synthesis

SMILES string

Cl.CCOC(=O)[C@@H](N)Cc1c[nH]c2ccccc12

InChI

1S/C13H16N2O2.ClH/c1-2-17-13(16)11(14)7-9-8-15-12-6-4-3-5-10(9)12;/h3-6,8,11,15H,2,7,14H2,1H3;1H/t11-;/m0./s1

InChI key

PESYCVVSLYSXAK-MERQFXBCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kento Takayama et al.
Journal of natural medicines, 75(1), 116-128 (2020-10-21)
Indole is produced from dietary tryptophan by tryptophanase in intestinal bacteria, such as Escherichia coli. In the liver, indole is converted into indoxyl sulfate, a uremic toxin and risk factor for chronic kidney disease (CKD). Probiotics and prebiotics are currently
Anikó Takátsy et al.
Journal of molecular recognition : JMR, 19(4), 270-274 (2006-05-17)
Studies of molecular recognition of chiral compounds by proteins are of importance from many points of view. The biological role of proteins in their interaction with small molecules is of fundamental interest and can be used in many different fields
Diana E Schlamadinger et al.
The journal of physical chemistry. B, 113(44), 14769-14778 (2009-10-13)
Ultraviolet resonance Raman (UVRR) spectra of tryptophan compounds in various solvents and a model peptide are presented and reveal systematic changes that reflect solvent polarity, hydrogen bond strength, and cation-pi interaction. The commonly utilized UVRR spectral marker for environment polarity
F L Tse et al.
The Journal of pharmacy and pharmacology, 36(9), 633-636 (1984-09-01)
Sandoz compound 57-118 is a mixture of tryptophan ethyl ester amide derivatives (analogues I-V) possessing one of five fatty acid chains which differ in chain length, configuration, or the degree of unsaturation. The relative absorption of each of the five
V Iu Shviadas et al.
Biokhimiia (Moscow, Russia), 45(4), 629-635 (1980-04-01)
The hydrolysis of L-tryptophane ethyl ester catalyzed by alpha-chymotrypsin and the effect of ethyl ster of D-tryptophane on the course of the reaction were studied. A kinetic pattern of a three-step enzymatic reaction based on the assumption that the enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service