Skip to Content
Merck
All Photos(2)

Key Documents

934046

Sigma-Aldrich

Ethyl 1,1,2,2-tetrafluoroethyl ether

greener alternative

≥99%, anhydrous, battery grade

Synonym(s):

1,1,2,2-Tetrafluoroethyl ethyl ether, 1-Ethoxy-1,1,2,2-tetrafluoroethane, ETFE, HFE 374

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6F4O
CAS Number:
Molecular Weight:
146.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

battery grade

Quality Level

Assay

≥99%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100 ppm HF
≤200 ppm H2O

bp

58 °C

mp

-86 °C (lit.)

density

1.21 g/mL

application(s)

battery manufacturing

greener alternative category

General description

Ethyl 1,1,2,2-tetrafluoroethyl ether (ETFE) is a fluorinated linear ether that has several key properties that make it useful in batteries. First, ETFE is miscible with many polar organic solvents used in electrolyte formulations, which improves the ionic conductivity of the battery. Additionally, ETFE has a relatively low viscosity (486 mPa•s at 23 °C) and a low boiling point (58 °C), which make it a versatile diluent for low-temperature applications. Our battery grade ETFE is manufactured with high purity and low trace impurities of water and acid.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Ethyl 1,1,2,2-tetrafluoroethyl ether (ETFE) is a highly effective additive in lithium-ion batteries and sodium-ion batteries for improving battery performance, increasing safety, and reducing flammability. ETFE is a versatile co-solvent with high miscibility, which allows it to be blended with other solvents to achieve high ionic conductivities. It is also a good SEI-forming electrolyte additive, stabilizing the Na electrode surface and enhancing the stability of high-voltage batteries. ETFE is particularly useful as an electrolyte co-solvent in lithium-sulfur (Li-S) batteries. Studies have shown that ETFE-containing electrolytes improve the stability of the electrolyte/anode interface and enhance the cycling, rate, and self-discharging of the Li-S cell more than glyme-based cosolvents or unblended solvents. With its unique properties and proven effectiveness, battery-grade ETFE is an excellent choice for manufacturers looking to create high-performing, reliable, and safe lithium-ion and sodium-ion batteries.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F

Flash Point(C)

-15 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nasim Azimi et al.
ACS applied materials & interfaces, 7(17), 9169-9177 (2015-04-14)
A high performance Li-S battery with novel fluoroether-based electrolyte was reported. The fluorinated electrolyte prevents the polysulfide shuttling effect and improves the Coulombic efficiency and capacity retention of the Li-S battery. Reversible redox reaction of the sulfur electrode in the
Synthesis and electrochemical properties of partially fluorinated ether solvents for lithium sulfur battery electrolytes,
Yue , et al.
Journal of Power Sources, 401, 271-277 (2018)
Lithium/sulfur batteries with mixed liquid electrolytes based on ethyl 1,1,2,2-tetrafluoroethyl ether,
Hai, et al.
Electrochimica Acta, 161, 55-62 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service