933473
Wafer tweezer
Style 3WL, 3" Handling
Synonym(s):
Wafer Tweezers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
EMS techware
Specificity
Handling 3", top lip, stepped bottom paddle.
Length: 120mm
Length: 120mm
Application
Wafer tweezer, Style 3WL, 3" are used for handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips. Made from antimagnetic/anti-acid steel with smooth surface.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service