Skip to Content
Merck
All Photos(1)

Key Documents

930105

Sigma-Aldrich

S-(4-nitrophenyl) methanethioate

≥95%

Synonym(s):

p-NO2-phenyl thioCORMate, p-NO2-phenyl thioformate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO3S
CAS Number:
Molecular Weight:
183.18
UNSPSC Code:
12352101
NACRES:
NA.06

Quality Level

Assay

≥95%

form

powder

reaction suitability

reaction type: C-C Bond Formation

functional group

nitro
thioether

storage temp.

2-8°C

SMILES string

[H]C(SC1=CC=C([N+]([O-])=O)C=C1)=O

Application

S-(4-nitrophenyl) methanethioate is an effective CO releasing molecule (CORM) developed by the lab of Sarah Tasker as part of a series of thioformate based CORMs called thioCORMates. p-NO2 phenyl thioformate generates CO rapidly for faster reactions that require instantaneous on-demand generation of CO. using only a simple amine to activate it.

This and the other thioCORMates are cost effective CORMs that when activated with simple amines, can be used to generate a near quantitative amount of CO safely without the usage of gas cylinders. These are compatible with a variety of carbonylation reactions, as well as existing COware glassware and components.

ThioCORMate: Tunable and Cost-Effective Carbon Monoxide-Releasing Molecule

Learn more about CO releasing molecules from Prof. Skrydstrup

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Tasker group's work focuses on the synthesis and utilization of stoichiometric CO-releasing molecules (CORMs) as alternatives to pressurized cylinders when the gas has safety, stability, or other practical handling concerns.

The Tasker group's work focuses on the synthesis and utilization of stoichiometric CO-releasing molecules (CORMs) as alternatives to pressurized cylinders when the gas has safety, stability, or other practical handling concerns.

The Tasker group's work focuses on the synthesis and utilization of stoichiometric CO-releasing molecules (CORMs) as alternatives to pressurized cylinders when the gas has safety, stability, or other practical handling concerns.

The Tasker group's work focuses on the synthesis and utilization of stoichiometric CO-releasing molecules (CORMs) as alternatives to pressurized cylinders when the gas has safety, stability, or other practical handling concerns.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service