Skip to Content
Merck
All Photos(1)

Key Documents

927449

Sigma-Aldrich

SDASO-L crosslinker

Synonym(s):

2,5-Dioxopyrrolidin-1-yl 3-((2-(3-(3-methyl-3H-diazirin-3-yl)propanamido)ethyl)sulfinyl) propanoate, Succinimidyl diazirine sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N4O6S
Molecular Weight:
372.40
UNSPSC Code:
12352106

form

chunks

Quality Level

storage temp.

−20°C

SMILES string

C1C(N(C(C1)=O)OC(CC[S](CCN(C(CCC2([C])N=N2)=O)[H])=O)=O)=O

Application

SDASO-L (succinimidyl diazirine sulfoxide, long version) is mass-spectrometry cleavable heterobifunctional cross-linker, which can be used in the analysis of protein structure, or protein-protein interactions. SDASO-L is photoactivated, allowing for the first time photocross-linking of proteins, and rapid, accurate analysis by MS.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Craig Gutierrez et al.
Molecular & cellular proteomics : MCP, 20, 100084-100084 (2021-04-30)
Cross-linking mass spectrometry (XL-MS) is a powerful tool for studying protein-protein interactions and elucidating architectures of protein complexes. While residue-specific XL-MS studies have been very successful, accessibility of interaction regions nontargetable by specific chemistries remain difficult. Photochemistry has shown great

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service