Skip to Content
Merck
All Photos(5)

Key Documents

926817

Sigma-Aldrich

Doped graphene oxide

greener alternative

nitrogen/sulfur co-doped, powder, bio-sourced

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303
NACRES:
NA.21

product name

Nitrogen/Sulfur co-doped graphene oxide powder, bio-sourced

description

Phase: Semi-crystalline

Quality Level

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

sheet size

, flake like structure

size

25-30 μm , particle size distribution method

surface area

7.7 m2/g , method- Nitrogen adsorption

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is produced in an environmental friendly way via bio-based processing. Click here for more information.

Application

This product is exclusively produced via bioprocess using earth friendly micro-organisms. No harmful chemicals or substances is used. Entire production process clean, sustainable, and economical. This product can be used as powder, dispersion in suitable solvents or liquid medium, and as composite for Chemical catalysis, Fuel cell catalysis, Semiconductor or Bio-chemical research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microbe-engaged synthesis of carbon dot decorated reduced graphene oxide as high performance oxygen reduction catalysts.
Zhou L, et al.
Journal of Material Chemistry A, 4, 7222-7222 (2016)
A one-pot biosynthesis of reduced graphene oxide (RGO)/bacterial cellulose (BC) nanocomposites.
Nandgaonkar A G, et al.
Green Chemistry, 16, 3195-3195 (2014)
Everett C Salas et al.
ACS nano, 4(8), 4852-4856 (2010-08-25)
Here we present that graphene oxide (GO) can act as a terminal electron acceptor for heterotrophic, metal-reducing, and environmental bacteria. The conductance and physical characteristics of bacterially converted graphene (BCG) are comparable to other forms of chemically converted graphene (CCG).

Articles

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service