Skip to Content
Merck
All Photos(1)

Key Documents

925985

Sigma-Aldrich

Niobium Aluminum Carbide MAX phase

Nb4AlC3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb4AlC3
Molecular Weight:
434.64
UNSPSC Code:
12352311
NACRES:
NA.54

Quality Level

form

powder

color

gray to black

solubility

insoluble

Application

Nb4AlC3 is the MAX phase precursor used for synthesis of Nb4C3Tx MXene, which have the below properties:

  • Electrical Conductivity: Single-layer Nb4C3Tx flakes boast an electrical conductivity of 1,024 ± 165 S cm−1.
  • Breakdown Current Density: ∼1.1 × 108 A.cm−2, comparable to Ti3C2Tx and graphene.
  • Electrochemical Performance: Specific capacity increases from 116 to 320 mAh.g−1 after 1,000 cycles at 1 A current density.
  • Interlayer Spacing: Accommodates ion intercalation/deintercalation without structural damage.
Unlock the potential of Nb4C3Tx MXene, using our Nb4AlC3 MAX precursor:
  • Electrochemical sensing: Nb4C3Tx excels in electrochemical sensing, detecting dopamine at 23 nm due to its high conductivity and enhanced intercalation.
  • Supercapacitors and Advanced batteries: Superior conductivity and charge-discharge rates for extended operation.
  • HER Catalyst: Efficient hydrogen evolution reactions.
  • ROS Scavengers: Biocompatible and photothermally stable for biomedical use.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prediction and Characterization of MXene Nanosheet Anodes for Non-Lithium-Ion Batteries
Xie, Yu et. al.
ACS Nano, 8, 9606?9615-9606?9615 (2014)
In Situ Reaction Synthesis, Electrical and Thermal, and Mechanical Properties of Nb4AlC3
Hu C, et al.
Journal of the American Chemical Society, 91, 2258-2263 (2008)
Recent advances in niobium MXenes: Synthesis, properties, and emerging applications
Rasheed, P. Abdul et. al.
Matter, 5(2), 546-572 (2022)

Articles

Optimizing the synthesis of high-quality 2D MXene flakes for 3D ink printing is essential to such technological developments as printable and flexible electronics.

Discover how MXenes' superior conductivity, strength, and stability are revolutionizing energy. Explore their potential for next-gen batteries and hydrogen evolution.

Discover how MXenes' superior conductivity, strength, and stability are revolutionizing energy. Explore their potential for next-gen batteries and hydrogen evolution.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service