Skip to Content
Merck
All Photos(1)

Key Documents

925500

Sigma-Aldrich

tBubpyCAMCN

≥95%

Synonym(s):

4,4′-Di-tert-butyl-2,2′-bipyridine-6-N-cyanocarboxamidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N5
Molecular Weight:
335.45
UNSPSC Code:
12352129
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

251-263 °C

storage temp.

−20°C

Application

tBubpyCAMCN is a bipyridine cyanocarboxamidine ligand that enables nickel-catalyzed cross-electrophile couplings of diverse aryl and alkyl halides.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service