Skip to Content
Merck
All Photos(1)

Key Documents

924903

Sigma-Aldrich

25-Hydroxyvitamin D2-(22,26,27-13C3) solution

50 μg/mL in ethanol, ≥98 atom % 13C, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C25H44O2
Molecular Weight:
415.63
UNSPSC Code:
12352005
NACRES:
NA.13

isotopic purity

≥98 atom % 13C

Quality Level

Assay

≥95% (CP)

concentration

50 μg/mL in ethanol

mass shift

M+3

SMILES string

C[C@@]12[C@](CC[C@]2([H])[C@H](C)/[13CH]=C/[C@H](C)C(O)([13CH3])[13CH3])([H])/C(CCC1)=C/C=C(C[C@@H](O)CC3)\C3=C

InChI key

KJKIIUAXZGLUND-DUOHQZGJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service