924091
Alkyne-a-TCO
Synonym(s):
(R,E)-1-(2-(Prop-2-yn-1-yloxy)ethyl)cyclooct-4-en-1-ol, 5-ax-Hydroxy-5-eq-(2-(prop-2-yn-1-yloxy)ethyl)-trans-cyclooctene, Clickable axial 5-hydroxy-trans-cyclooctene
Select a Size
€254.00
Availability
Select a Size
About This Item
€254.00
Availability
reaction suitability
reagent type: chemical modification reagent
reagent type: cross-linking reagent
reaction type: click chemistry
Quality Level
functional group
(Alkyne)
storage temp.
−20°C
Application
Other new a-TCOs:
- NHS ester-a-TCO (924075)
- TAMRA-a-TCO (924083)
- Alkyne-a-TCO (924091)
- Amine-a-TCO (924113)
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service