923389
1-((2-(tert-Butoxycarbonyl)-5-oxa-2-azaspiro[3.4]octan-6-yl)methyl)piperidine-4-carboxylic acid
Synonym(s):
Rigid linker for PROTAC® development
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H30N2O5
Molecular Weight:
354.44
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22
Recommended Products
form
powder or crystals
Quality Level
reaction suitability
reagent type: linker
functional group
Boc
carboxylic acid
storage temp.
2-8°C
SMILES string
O=C(O)C(CC1)CCN1CC(CC2)OC32CN(C(OC(C)(C)C)=O)C3
InChI key
CVHPZSHLWZEYQS-UHFFFAOYSA-N
Related Categories
Application
1-((2-(tert-Butoxycarbonyl)-5-oxa-2-azaspiro[3.4]octan-6-yl)methyl)piperidine-4-carboxylic acid is useful as a rigid linker in PROTAC development for targeted protein degradation as well as chemical conjugates. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties. Developed in partnership with ComInnex.
Other Notes
Legal Information
Practum is a registered trademark of Sartorius Lab Instruments GmbH & Co. KG
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service