Skip to Content
Merck
All Photos(1)

Key Documents

920819

Sigma-Aldrich

mPEG functionalized alginate

5% +/- 2% PEGylation, PEG average Mn 1k

Synonym(s):

PEGylated alginate, PEGylated low viscosity alginate, PEGylated sodium alginate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
{C6H7O5[NHCH2CH2CH2O(CH2CH2O)xCH3]}n(C6H7O6Na)m
UNSPSC Code:
12352201
NACRES:
NA.23

Quality Level

form

(solid or chunk(s) or fiber)

mol wt

PEG average Mn 1k

color

white to off-white

storage temp.

2-8°C

Application

Alginate is biocompatible and biodegradable, and forms hydrogels in the presence of divalent cations (e.g., Ca2+, Ba2+) under mild temperature and pH conditions. mPEG functionalized alginate can used to tune the stress relaxation of alginate hydrogels, and can used to fabricate microparticles with better mechanical properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solène Passemard et al.
Biomacromolecules, 18(9), 2747-2755 (2017-07-26)
The production of hydrogel microspheres (MS) for cell immobilization, maintaining the favorable properties of alginate gels but presenting enhanced performance in terms of in vivo durability and physical properties, is desirable to extend the therapeutic potential of cell transplantation. A
Varying PEG density to control stress relaxation in alginate-PEG hydrogels for 3D cell culture studies
Nam S., et al.
Biomaterials, 200, 15-24 (2019)
Redouan Mahou et al.
Materials (Basel, Switzerland), 7(1), 275-286 (2014-01-09)
The progress of medical therapies, which rely on the transplantation of microencapsulated living cells, depends on the quality of the encapsulating material. Such material has to be biocompatible, and the microencapsulation process must be simple and not harm the cells.
Xu Jia et al.
ACS biomaterials science & engineering, 2(9), 1641-1648 (2016-09-12)
Theranostic nanomedicine has recently emerged as an appealing approach for tumor chemotherapy. Here, for the first time, fluorescent carbon dots (Cdots) were used as cross-linker for tumor theranostic nanoparticles. Novel theranostic nanoparticles of approximately 27 nm with a doxorubicin (DOX)
Sungmin Nam et al.
Biomaterials, 200, 15-24 (2019-02-12)
Hydrogels are commonly used as artificial extracellular matrices for 3D cell culture and for tissue engineering. Viscoelastic hydrogels with tunable stress relaxation have recently been developed, and stress relaxation in the hydrogels has been found to play a key role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service