All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C40H32N10Ni
CAS Number:
Molecular Weight:
711.44
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
form
crystals
reaction suitability
reagent type: catalyst
reaction type: Cross Couplings
mp
>300 °C
SMILES string
[Ni+2].[N-]5\C(=N\c8nccc(c8)C)\c6c(cccc6)\C\5=N/c7nccc(c7)C.[N-]1\C(=N\c4nccc(c4)C)\c2c(cccc2)\C\1=N/c3nccc(c3)C
InChI key
UEBWUPHCLCWDLU-UHFFFAOYSA-N
Related Categories
Application
(MeBPI)2Ni is a redox-active shuttle developed in the Sevov lab for electrocatalytic cross-electrophile coupling reactions. When combined with (MeBPI)Ni-OAc (919365), (MeBPI)Ni-OAc enables the reductive coupling of a wide range of aryl, heteroaryl, or vinyl bromides with primary or secondary alkyl bromides.
Product can be used with our Synlectro electrochemistry platform (ESYNTH001)
Product can be used with our Synlectro electrochemistry platform (ESYNTH001)
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Blaise L Truesdell et al.
Journal of the American Chemical Society, 142(12), 5884-5893 (2020-03-03)
Cross-electrophile coupling (XEC) of alkyl and aryl halides promoted by electrochemistry represents an attractive alternative to conventional methods that require stoichiometric quantities of high-energy reductants. Most importantly, electroreduction can readily exceed the reducing potentials of chemical reductants to activate catalysts
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service