918008
GPhos
≥95%
Synonym(s):
(3-(tert-Butoxy)-2′,6′-diisopropyl-6-methoxy-[1,1′-biphenyl]-2-yl)dicyclohexylphosphane
About This Item
Recommended Products
Quality Level
Assay
≥95%
form
powder
reaction suitability
reagent type: catalyst
reaction type: Cross Couplings
reagent type: ligand
greener alternative product score
old score: 16
new score: 13
Find out more about DOZN™ Scoring
greener alternative product characteristics
Designing Safer Chemicals
Safer Solvents and Auxiliaries
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
mp
230-233 °C
functional group
phosphine
greener alternative category
General description
Application
Learn more about Buchwald Phosphine Ligands
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service