Skip to Content
Merck
All Photos(1)

Key Documents

915777

Sigma-Aldrich

1-Ethyl-3-methylimidazolium tetracyanoborate Solarpur®

Synonym(s):

EMIM TCB Solarpur®, [EMIM][B(CN)4]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11BN6
CAS Number:
Molecular Weight:
226.05
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥99.5% (HPLC)

Quality Level

form

liquid

impurities

≤100 ppm Halides
≤100 ppm Water

SMILES string

[B-](C#N)(C#N)(C#N)C#N.[n+]1(c[n](cc1)C)CC

InChI

1S/C6H11N2.C4BN4/c1-3-8-5-4-7(2)6-8;6-1-5(2-7,3-8)4-9/h4-6H,3H2,1-2H3;/q+1;-1

InChI key

CPZCMVDICTVHIP-UHFFFAOYSA-N

Application

1-Ethyl-3-methylimidazolium tetracyanoborate can be utilized as an electrolyte or component in the electrolyte formulation of dye-sensitized solar cells. Ionic liquids, including1-Ethyl-3-methylimidazolium tetracyanoborate, can be used as components in advanced energy storage systems such as supercapacitors or batteries.
EMIM TCB Solarpur® is high-purity, low friction electronic grade ionic liquid electrolyte for various applications including transparent electrodes for stretchable electronics, DSSCs, and gas separation membranes.

Solarpur® electrolyte components for DSSC applications meet the highest purity standards regarding water and other impurities required for this technology.

Legal Information

Solarpur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced Efficiency of Dye-Sensitized Solar Cells with Mesoporous-Macroporous TiO2 Photoanode Obtained Using ZnO Template.
Pham T T T, et al.
Journal of Electronic Materials, 46(6), 3801-3807 (2017)
Mei Ying Teo et al.
ACS applied materials & interfaces, 9(1), 819-826 (2016-12-20)
Stretchable conductive materials have received great attention owing to their potential for realizing next-generation stretchable electronics. However, the simultaneous achievement of excellent mechanical stretchability and high electrical conductivity as well as cost-effective fabrication has been a significant challenge. Here, we
Mingshi Jin et al.
Journal of nanoscience and nanotechnology, 12(1), 815-821 (2012-04-25)
The light harvesting efficiency of dye-sensitized solar cells was enhanced by using a scattering layer. Such as sphere type TiO2, inverse photonic crystal TiO2, hollow spherical TiO2. Among these materials, the TiO2 with inverse photonic crystal (IPC) structure, synthesized by
Scalable application of thin film coating techniques for supported liquid membranes for gas separation made from ionic liquids.
Gruenauer J, et al.
Journal of Membrane Science , 518, 178-191 (2016)
Seyoung Kee et al.
Advanced materials (Deerfield Beach, Fla.), 30(3) (2017-12-07)
Despite the high expectation of deformable and see-through displays for future ubiquitous society, current light-emitting diodes (LEDs) fail to meet the desired mechanical and optical properties, mainly because of the fragile transparent conducting oxides and opaque metal electrodes. Here, by

Articles

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service