Skip to Content
Merck
All Photos(3)

Key Documents

915610

Sigma-Aldrich

PrPPTNO

≥95%

Synonym(s):

2,4,6,8-Tetraoxo-1,3,7,9-tetrapropyl-1,2,3,4,6,7,8,9-octahydropyrimido[5,4-g]pteridine 5-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N6O5
CAS Number:
Molecular Weight:
432.47
UNSPSC Code:
12352101

Assay

≥95%

form

powder

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

Application

PrPPTNO is an organic photocatalyst developed in the Pospech group for the E/Z-isomerization of stilbenes as well as the oxidative cyclization of 2-phenyl benzoic acids.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richy Hauptmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(17), 4325-4329 (2019-02-07)
Herein we report the photo- and electrochemical characterization of pyrimidopteridine N-oxide-based heterocycles. The potential of their application as organic photoredox catalysts is showcased in the photomediated contra-thermodynamic E→Z isomerization of cinnamic acid derivatives and oxidative cyclization of 2-phenyl benzoic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service