915319
Aminosupersilane
Synonym(s):
N-(Adamantan-1-yl)-1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilan-2-amine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C19H43NSi4
Molecular Weight:
397.89
UNSPSC Code:
12352116
NACRES:
NA.06
Recommended Products
form
powder
Quality Level
Related Categories
Application
Aminosupersilane has been demonstrated by the MacMillan lab to be a necessary reagent in the dual catalytic nickel/photoredox cross-electrophile coupling of unactivated alkyl chlorides. The increased nucleophilicity of aminosupersilane, as compared to other organosilane reagents, is key to activating the alkyl chloride for chlorine atom abstraction.
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service