Skip to Content
Merck
All Photos(2)

Key Documents

914797

Sigma-Aldrich

Birch O-PC C0103

≥97%, New Iridium

Synonym(s):

2-(2-Ethylhexyl)-6,8,11-tris(4-methoxyphenyl)-1H-peryleno[1,12-efg]isoindole-1,3(2H)-dione, BPI-p-OMePh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H45NO5
Molecular Weight:
775.93
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥97%

form

powder

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

Application

Birch O-PC C0103 is an organic photocatalyst developed in the Miyake lab for the photocatalytic Birch reduction driven by visible light. The hallmark high excited state redox potential of this first of its kind catalyst class reduces benzene and other arenes to produce functionally useful 1,4-cyclohexadienes. This organic photocatalyst accesses both classical and non-classical Birch reduction reactivity in addition to demonstrating hydrodechlorination of aromatics. Employing mild benchtop conditions, this photocatalytic approach eliminates challenges associated with traditional Birch reduction such as the use of alkali metals in liquid ammonia.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Legal Information

Sold in collaboration with New Iridium LLC. For orders greater than 25g, please contact New Iridium at chern@newiridium.com or visit https://www.newiridium.com.
Birch O-PC is a trademark of New Iridium Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service