Skip to Content
Merck
All Photos(2)

Key Documents

910945

Sigma-Aldrich

di-phenylethylammonium lead(II) tetrabromide

greener alternative

n = 1

Synonym(s):

(PEA)2PbBr4, 2D perovskite, Bis[(2-phenylethyl)ammonium] tetrabromoplumbate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2PbBr4
CAS Number:
Molecular Weight:
771.19
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[Pb](Br)(Br)(Br)Br.[N+H3]CCc2ccccc2.[N+H3]CCc1ccccc1

InChI key

SLXREHWTSXNZOP-UHFFFAOYSA-L

General description

This product is a violet emmiter for light emmiting diodes. It is solution processable in DMF to create thin films. This material has a photoluminescence peak at 407 nm with a quantum yield of 26% as nanoplatelets. Single crystals have been grown with a 412 nm emission of and quantum efficiency of 0.22(1). LED devices using this material as the emissive layer have been constructed that opperate at room temperature with an on/off voltage of 2.5-2.8 V and external quantum efficiency (EQE) of 0.038%.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for "Energy efficiency".

Application

di-phenylethylammonium lead(II) tetrabromide can be used in the field of solar cells and photovoltaic devices as a precursor material for the synthesis of lead halide perovskite thin films, which have shown promising efficiency in solar energy conversion. di-phenylethylammonium lead(II) tetrabromide based perovskite materials have been studied for their potential in optoelectronic devices such as light-emitting diodes (LEDs), lasers and photodetectors.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrathin Two-Dimensional Organic?Inorganic Hybrid Perovskite Nanosheets with Bright, Tunable Photoluminescence and High Stability
Yang S, et al.
Angewandte Chemie (International Edition in English), 56, 4252-4252 (2017)
Color-Pure Violet-Light-Emitting Diodes Based on Layered Lead Halide Perovskite Nanoplates
Liang D, et al.
ACS Nano, 10, 6897-6897 (2016)
Giant Two-Photon Absorption and Its Saturation in 2D Organic?Inorganic Perovskite
Li W, et al.
Advanced Optical Materials, 5, 1601045-1601045 (2017)
Giant Two-Photon Absorption and Its Saturation in 2D Organic?Inorganic Perovskite.
Liu W, et al.
Advanced Optical Materials, 5, 1601045-1601045 (2017)
Effects of Organic Moieties on Luminescence Properties of Organic-Inorganic Layered Perovskite-Type Compounds
Kawano N, et al.
The Journal of Physical Chemistry C, 118, 9101-9101 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service