Skip to Content
Merck
All Photos(1)

Key Documents

907375

Sigma-Aldrich

H-L-Photo-methionine HCl

≥95%

Synonym(s):

(S)-2-Amino-4-(3-methyl-3H-diazirin-3-yl)butanoic acid hydrochloride, (S)-2-Amino-4-(3H-diazirin-3-yl)pentanoic acid hydrochloride, Diazirine amino acid, Photo-Met, Photo-crosslinking amino acid, Photoprobe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12ClN3O2
CAS Number:
Molecular Weight:
193.63
UNSPSC Code:
12352209

Assay

≥95%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

availability

available only in USA

application(s)

peptide synthesis

storage temp.

2-8°C

Application

H-L-Photo-Methionine HCl is a diazirine-containing methionine amino acid and multifunctional photo-crosslinker. Its incorporation into peptides or small-molecule probes and tools allows for photoaffinity labeling of cellular targets and protein-protein interactions upon UV light (~360 nm) irradiation to form a covalent bond. This and other multifunctional probe building blocks will continue to accelerate drug discovery research for probing cellular mechanisms, target ID/validation, and understanding traditionally undruggable targets. An Fmoc-protected version is also available as 907367.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeolin Lee et al.
Analytical chemistry, 88(19), 9503-9509 (2016-09-01)
Fc-specific antibody binding proteins (FcBPs) with the minimal domain of protein G are widely used for immobilization of well-oriented antibodies onto solid surfaces, but the noncovalently bound antibodies to FcBPs are unstable in sera containing large amounts of antibodies. Here
Xiaozhou Luo et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(13), 3615-3620 (2016-03-16)
Thiopeptides are a subclass of ribosomally synthesized and posttranslationally modified peptides (RiPPs) with complex molecular architectures and an array of biological activities, including potent antimicrobial activity. Here we report the generation of thiopeptides containing noncanonical amino acids (ncAAs) by introducing
Claudio Iacobucci et al.
Analytical chemistry, 90(4), 2805-2809 (2018-01-30)
A major challenge in cross-linking/mass spectrometry (MS) is targeting carboxyl functions in proteins under physiological conditions that do not disturb the protein's conformation. Cross-linking of glutamic acid and aspartic acid residues in proteins will greatly expand the scope of structural
Michael J Bollong et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(46), E9903-E9912 (2017-11-01)
Expression of the transcription factor FOXC2 is induced and necessary for successful epithelial-mesenchymal transition, a developmental program that when activated in cancer endows cells with metastatic potential and the properties of stem cells. As such, identifying agents that inhibit the
Haibin Shi et al.
Journal of the American Chemical Society, 134(6), 3001-3014 (2012-01-17)
Protein kinases (PKs) play an important role in the development and progression of cancer by regulating cell growth, survival, invasion, metastasis, and angiogenesis. Dasatinib (BMS-354825), a dual Src/Abl inhibitor, is a promising therapeutic agent with oral bioavailability. It has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service