Skip to Content
Merck
All Photos(1)

Key Documents

906018

Sigma-Aldrich

Spiro[9H-fluorene-9,9′-[9H]xanthene]-2,2′,7,7′-tetramine

Synonym(s):

N2,N2,N7,N7,N2′,N2′,N7′,N7′-octakis(4-methoxyphenyl)spiro[9H-fluorene-9,9′-[9H]xanthene]-2,2′,7,7′-tetramine, X60

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C81H68N4O9
CAS Number:
Molecular Weight:
1241.43
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.23

Assay

>98%

form

solid

color

beige

SMILES string

COC1=CC=C(C=C1)N(C2=CC=C(OC)C=C2)C3=CC=C4C(C5(C6=CC(N(C7=CC=C(OC)C=C7)C8=CC=C(OC)C=C8)=CC=C6OC9=CC=C(N(C%10=CC=C(OC)C=C%10)C%11=CC=C(OC)C=C%11)C=C59)C%12=CC(N(C%13=CC=C(OC)C=C%13)C%14=CC=C(OC)C=C%14)=CC=C4%12)=C3

InChI

1S/C81H68N4O9/c1-86-65-29-9-53(10-30-65)82(54-11-31-66(87-2)32-12-54)61-25-45-73-74-46-26-62(83(55-13-33-67(88-3)34-14-55)56-15-35-68(89-4)36-16-56)50-76(74)81(75(73)49-61)77-51-63(84(57-17-37-69(90-5)38-18-57)58-19-39-70(91-6)40-20-58)27-47-79(77)94-80-4

InChI key

BSBVJQUURRFRGT-UHFFFAOYSA-N

General description

Spiro[9H-fluorene-9,9′-[9H]xanthene]-2,2′,7,7′-tetramine (X60) is a hole transporting material (HTM) with a nonplanar 3D structure which reduces the electronic coupling and charge recombination. It can be used as a starting material in the fabrication of green and cost effective p-type organic semiconductors.
X60 is cheaper alternative to Spiro-OMeTAD.
X60 is a spiro[fluorene-9,9′-xanthene] (SFX) based organic hole transport material (HTM).
Devices with X60 as HTM showed high power conversion efficiencies (PCEs) amounting to 7.30% in solid-state dye-sensitized solar cells (ssDSCs) and 19.84% in perovskite solar cells (PSCs). These results are competitive with the reported record PCEs of ssDSC and PSC devices based on the well-known HTM Spiro-OMeTAD. In addition, almost no hysteretic behavior could be observed in the X60-based PSCs, and the photovoltaic parameters of the devices are not affected by the scan rates used.

Application

X60 can be used as a conducting polymer in the formation of perovskite based solar cells with high efficiency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SFX as a low-cost ′Spiro′ hole-transport material for efficient perovskite solar cells.
Maciejczyk M, et al.
Journal of Material Chemistry A, 4(13), 4855-4863 (2016)
Precise Control of Thermal and Redox Properties of Organic Hole-Transport Materials
Chiykowski VA, et al.
Angewandte Chemie (International Edition in English), 130(47), 15755-15759 (2018)
A low-cost spiro[fluorene-9,90-xanthene]-based hole transport material for highly efficient solid-state dye-sensitized solar cells and perovskite solar cells.
Xu B, et al.
Energy & Environmental Science, 9(3), 873-877 (2016)
Effects of different small molecule hole transporters on the performance and charge transfer dynamics of perovskite solar cells
Pydzinska K, et al.
Synthetic Metals, 232(47), 181-187 (2017)
Building Solar Cells from Nanocrystal Inks
Luo K, et al.
Applied Sciences, 9(9), 1885-1885 (2019)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service