Skip to Content
Merck
All Photos(1)

Key Documents

905062

Sigma-Aldrich

Perovskite quantum dots

oleic acid and oleylamine coated, fluorescence λem 530 nm, 10 mg/mL in toluene

Synonym(s):

Cadmium free QDs, Fluorescent nanocrystals, Perovskite FAPbX3 Quantum Dots, Perovskite nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH5N2PbBr3
Molecular Weight:
491.98
UNSPSC Code:
12352300
NACRES:
NA.23

form

liquid

concentration

10 mg/mL in toluene

fluorescence

λem 530 nm

storage temp.

2-8°C

General description

Perovskite quantum dots (PQDs) are semiconducting materials with high luminescent efficiency. They have a low threshold, tuneable wavelength and ultra-stable stimulated emission (SE). These semiconductors are a class of hybrid inorganic and organic metal halide based perovskite materials. They have a direct bandgap which can be useful for a variety of optoelectronic devices.

Application

PQDs can be used in the fabrication of light emitting diodes (LEDs) and PQD based solar cells.
Perovskite quantum dots (QDs) have the common formula ABX3 where A is Cs or FA (formamidinium), X is Cl, Br or I. They possess high photoluminescence efficiency and narrow emission and emit in the visible spectral regime. Perovskite QDs are cadmium free and the aforementioned properties render them suitable for applications in light emitting diodes (LEDs), lasers, liquid crystal displays (LCDs) etc.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

All-inorganic colloidal perovskite quantum dots: a new class of lasing materials with favorable characteristics
Wang Y, et al.
Advanced Materials, 27(44), 7101-7108 (2015)
Nanocrystals of Cesium Lead Halide Perovskites (CsPbX3, X = Cl, Br, and I): Novel Optoelectronic Materials Showing Bright Emission with Wide Color Gamut
L Protesescu et al.
Nano Letters, 15, 3692-3692 (2015)
Brightly luminescent and color-tunable formamidinium lead halide perovskite FAPbX3 (X= Cl, Br, I) colloidal nanocrystals
Levchuk I, et al.
Nano Letters, 17(5), 2765-2770 (2017)
Multicolored Mixed-Organic-Cation Perovskite Quantum Dots (FA x MA1-x PbX3, X= Br and I) for White Light-Emitting Diodes
Zhang Y, et al.
Industrial & Engineering Chemistry Research, 56(36), 10053-10059 (2017)
CsPbX3 quantum dots for lighting and displays: room-temperature synthesis, photoluminescence superiorities, underlying origins and white light-emitting diodes
Li X, et al.
Advances in Functional Materials, 26(15), 2435-2445 (2016)

Articles

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service